Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133798.3 | NP_598559.2 | 3, 4 | 909 | |
GenBank mRNA | AK146901.1 | - | 3, 4 | 790 | |
AK147922.1 | - | 3, 4 | 770 | ||
AK158201.1 | - | 3, 4 | 800 | ||
AK162206.1 | - | 2, 3 | 677 | ||
AK164329.1 | - | 3, 4 | 785 | ||
AK167692.1 | - | 3, 4 | 910 | ||
BC018508.1 | - | 2, 3 | 452 | 3156447 |