Gene Transcripts
- Gene Symbol
- Rars
- Entrez Gene ID
- 104458
- Gene Name
- arginyl-tRNA synthetase
- Gene Aliases
- 2610011N19Rik, 2610037E21Rik, AL033339, AW985894
- Chromosome Location
- Chr. 11: 35808381 - 35834528 on Build GRCm38
- UniGene ID
- Mm.284906
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025936.3 |
NP_080212.2
|
7, 8 |
862 |
|
GenBank mRNA |
AK011383.1 |
- |
7, 8 |
840 |
|
AK076160.1 |
- |
7, 8 |
837 |
|
AK149856.1 |
- |
7, 8 |
837 |
|
AK151168.1 |
- |
1, 2 |
28 |
|
AK151966.1 |
- |
6, 7 |
660 |
|
AK152108.1 |
- |
7, 8 |
837 |
|
AK160216.1 |
- |
7, 8 |
837 |
|
AK166097.1 |
- |
7, 8 |
837 |
|
AK168194.1 |
- |
7, 8 |
837 |
|
BC020132.1 |
- |
7, 8 |
820 |
3989952 |
View Full Product Details