Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014976.2 | NP_001014976.1 | 16 | 3166 | |
GenBank mRNA | AK129072.1 | - | 16 | 3133 | |
AY588413.1 | - | 16 | 3104 | ||
BC082603.1 | - | 16 | 3166 | 30668743 | |
BC145844.1 | - | 16 | 3137 | 40131147 | |
BC145846.1 | - | 16 | 3083 | 40131149 |