Gene Transcripts

Gene Symbol
ORM1
Entrez Gene ID
5004
Gene Name
orosomucoid 1
Gene Aliases
AGP-A, AGP1, HEL-S-153w, ORM
Chromosome Location
Chr. 9: 114323023 - 114326479 on Build GRCh38
UniGene ID
Hs.522356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000607.2 NP_000598.2 4 511
GenBank mRNA AK312035.1 - 4 436
BC026238.1 - 4 436 4767956
BC104818.1 - 4 425 8143821
BC104820.1 - 4 425 8143823
BC143313.1 - 4 425 9051818
BC143314.1 - 4 425 9051819
BI908589.1 - 4 510
BT019790.1 - 4 400
EU794584.1 - 4 400
M13692.1 - 4 415
X02544.1 - 4 478

View Full Product Details