Gene Transcripts

Gene Symbol
Chn1
Entrez Gene ID
108699
Gene Name
chimerin 1
Gene Aliases
0610007I19Rik, 0710001E19Rik, 1700112L09Rik, 2900046J01Rik, AI413815, ARHGAP2
Chromosome Location
Chr. 2: 73610660 - 73775367 on Build GRCm38
UniGene ID
Mm.257073
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113246.2 NP_001106717.2 9, 10 1194
NM_029716.3 NP_083992.1 2, 3 395
NM_175752.3 NP_786928.2 3, 4 1006
GenBank mRNA AB264774.1 - 1, 2 796
AF332069.1 - 3, 4 514
AF332070.1 - 3, 4 514
AK007182.1 - 2, 3 395
AK032630.1 - 3, 4 964
AK049943.1 - 3, 4 950
AK075606.1 - 3, 4 968
AK147686.1 - 3, 4 950
BC010825.1 - 3, 4 981 4219273
BC024796.1 - 3, 4 957 5360438
BC025023.1 - 3, 4 937 5364554
BC054770.1 - 9, 10 946 6415204

View Full Product Details