Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027288.3 | NP_081564.3 | 14 | 2003 | |
GenBank mRNA | AF306557.1 | - | 14 | 2007 | |
AK142165.1 | - | 14 | 1968 | ||
AK154172.1 | - | 14 | 1985 | ||
AK166022.1 | - | 14 | 1986 | ||
AK166340.1 | - | 14 | 2001 | ||
AK170290.1 | - | 14 | 2000 | ||
BC031409.1 | - | 14 | 1946 | 4952710 |