Gene Transcripts

Gene Symbol
ZAR1
Entrez Gene ID
326340
Gene Name
zygote arrest 1
Gene Aliases
Z3CXXC6
Chromosome Location
Chr. 4: 48490252 - 48497528 on Build GRCh38
UniGene ID
Hs.276457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175619.2 NP_783318.1 2 1020
GenBank mRNA AY191416.1 - 2 980
BC136265.1 - 2 1020 9020252

View Full Product Details