Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001255.2 | NP_001246.2 | 5 | 600 | |
GenBank mRNA | AF099644.1 | - | 4 | 543 | |
AK225504.1 | - | 5 | 579 | ||
AK302539.1 | - | 3 | 832 | ||
AK303844.1 | - | 6 | 555 | ||
AK312780.1 | - | 5 | 579 | ||
BC000624.2 | - | 4 | 607 | 3346682 | |
BC001088.2 | - | 5 | 600 | 2964686 | |
BC006272.2 | - | 4 | 542 | 3948819 | |
BC009425.1 | - | 5 | 582 | 2959460 | |
BC009426.1 | - | 5 | 582 | 2959596 | |
BC010044.2 | - | 5 | 591 | 3634656 | |
BC012803.2 | - | 5 | 582 | 2959588 | |
BC012827.2 | - | 5 | 600 | 2964686 | |
BC013303.2 | - | 5 | 582 | 2959588 | |
BC015998.1 | - | 5 | 578 | 4306426 | |
BC024257.1 | - | 5 | 582 | 2959460 | |
BC031294.1 | - | 5 | 582 | 2959487 | |
BC110321.1 | - | 4 | 563 | 5496957 | |
BT007388.1 | - | 4 | 499 | ||
U05340.1 | - | 5 | 609 |