Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320595.1 | NP_001307524.1 | 4 | 651 | |
NM_001320596.1 | NP_001307525.1 | 4 | 540 | ||
NM_001320597.1 | NP_001307526.1 | 3 | 391 | ||
NM_004396.4 | NP_004387.1 | 3 | 626 | ||
GenBank mRNA | AB209919.1 | - | 3 | 378 | |
AB451257.1 | - | 3 | 224 | ||
AB451382.1 | - | 3 | 224 | ||
AK297192.1 | - | 3 | 393 | ||
AK297753.1 | - | 4 | 361 | ||
AK310318.1 | - | 3 | 270 | ||
BC016027.1 | - | 3 | 365 | 3528578 | |
BT006943.1 | - | 3 | 224 | ||
BX571764.1 | - | 3 | 395 | ||
DA191631.1 | - | 3 | 471 | ||
DB231101.1 | - | 3 | 391 | ||
X15729.1 | - | 3 | 391 | ||
X52104.1 | - | 3 | 399 |