Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020675.3 | NP_065726.1 | 3 | 305 | |
XM_011511516.2 | XP_011509818.1 | 3 | 314 | ||
GenBank mRNA | AF225416.1 | - | 3 | 287 | |
AK291093.1 | - | 3 | 316 | ||
AY456388.1 | - | 2 | 174 | ||
BC022255.1 | - | 3 | 305 | 4738187 | |
CR457197.1 | - | 2 | 174 |