Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020675.3 | NP_065726.1 | 7 | 715 | |
GenBank mRNA | AF225416.1 | - | 7 | 697 | |
AK291093.1 | - | 7 | 726 | ||
AY456388.1 | - | 6 | 584 | ||
BC022255.1 | - | 7 | 715 | 4738187 | |
CR457197.1 | - | 6 | 584 |