Gene Transcripts

Gene Symbol
IFNGR2
Entrez Gene ID
3460
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Gene Aliases
AF-1, IFGR2, IFNGT1, IMD28
Chromosome Location
Chr. 21: 33402896 - 33437521 on Build GRCh38
UniGene ID
Hs.634632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005534.3 NP_005525.2 7 1910
XM_005260969.2 XP_005261026.1 8 1426
XM_011529553.1 XP_011527855.1 8 1352
XM_011529554.2 XP_011527856.1 7 1786
GenBank mRNA BC003624.2 - 7 1377
U05875.1 - 7 1911
U05877.1 - 7 1375

View Full Product Details