Gene Transcripts

Gene Symbol
Tmtc1
Entrez Gene ID
387314
Gene Name
transmembrane and tetratricopeptide repeat containing 1
Gene Aliases
Arg99, BC023818
Chromosome Location
Chr. 6: 148232430 - 148444845 on Build GRCm38
UniGene ID
Mm.137858
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198967.5 NP_945318.2 20 3897
GenBank mRNA AK046396.1 - 19 3786
AK137545.1 - 20 3867
BC023818.1 - 9 2058

View Full Product Details