Gene Transcripts

Gene Symbol
Zmynd10
Entrez Gene ID
114602
Gene Name
zinc finger, MYND domain containing 10
Gene Aliases
Blu
Chromosome Location
Chr. 9: 107545991 - 107551321 on Build GRCm38
UniGene ID
Mm.28331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053253.3 NP_444483.2 9, 10 1139
GenBank mRNA AF123386.1 - 7, 8 770
AK132332.1 - 9, 10 1148
BC052357.1 - 9, 10 1139 6772144

View Full Product Details