Gene Transcripts

Gene Symbol
Snrpg
Entrez Gene ID
68011
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
2810024K17Rik, AL022803, SMG, sm-G, snRNP-G
Chromosome Location
Chr. 6: 86371540 - 86378902 on Build GRCm38
UniGene ID
Mm.276802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026506.2 NP_080782.1 3 168
GenBank mRNA AK011129.1 - 3 167
AK012802.1 - 3 157
AK013235.1 - 3 169
AK017852.1 - 3 517
BC027499.1 - 3 141 1430048
BC051470.1 - 3 213 1244608
BC094411.1 - 3 155 1380524

View Full Product Details