Gene Transcripts
- Gene Symbol
- Coro1a
- Entrez Gene ID
- 155151
- Gene Name
- coronin, actin binding protein 1A
- Gene Aliases
- Chromosome Location
- Chr. 1: 198123883 - 198128857 on Build Rnor_6.0
- UniGene ID
- Rn.6990
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130411.2 |
NP_569095.1
|
5, 6 |
734 |
|
XM_006230204.2 |
XP_006230266.1
|
5, 6 |
818 |
|
GenBank mRNA |
AF416730.1 |
- |
4, 5 |
630 |
|
AF495469.1 |
- |
5, 6 |
734 |
|
BC086971.1 |
- |
5, 6 |
710 |
7124429 |
FQ225277.1 |
- |
2, 3 |
256 |
|
FQ225360.1 |
- |
2, 3 |
256 |
|
FQ225522.1 |
- |
1, 2 |
172 |
|
FQ226290.1 |
- |
3, 4 |
203 |
|
FQ226379.1 |
- |
5, 6 |
722 |
|
FQ226941.1 |
- |
5, 6 |
754 |
|
FQ233306.1 |
- |
5, 6 |
728 |
|
FQ233390.1 |
- |
5, 6 |
730 |
|
FQ233414.1 |
- |
5, 6 |
732 |
|
FQ233980.1 |
- |
5, 6 |
647 |
|
FQ234750.1 |
- |
5, 6 |
733 |
|
View Full Product Details