Gene Transcripts

Gene Symbol
Slc24a2
Entrez Gene ID
84550
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 2
Gene Aliases
Nckx2
Chromosome Location
Chr. 5: 105336287 - 105579959 on Build Rnor_6.0
UniGene ID
Rn.208755
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031743.2 NP_113931.1 1 261
GenBank mRNA AF021923.1 - 1 261

View Full Product Details