Gene Transcripts

Gene Symbol
Kif5c
Entrez Gene ID
16574
Gene Name
kinesin family member 5C
Gene Aliases
KINN, Khc, NKHC, NKHC2
Chromosome Location
Chr. 2: 49619314 - 49774778 on Build GRCm38
UniGene ID
Mm.256342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008449.2 NP_032475.2 26 6397
GenBank mRNA AB093244.1 - 26 6289
AK134577.1 - 1 627
BC067051.1 - 26 6397 30536079

View Full Product Details