Gene Transcripts

Gene Symbol
CCDC144NL
Entrez Gene ID
339184
Gene Name
coiled-coil domain containing 144 family, N-terminal like
Gene Aliases
Chromosome Location
Chr. 17: 20847348 - 20896587 on Build GRCh38
UniGene ID
Hs.674830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004306.2 NP_001004306.1 4 1000
GenBank mRNA BC068597.1 - 4 1000

View Full Product Details