Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083961.1 | NP_001077430.1 | 17 | 2194 | |
NM_173636.4 | NP_775907.4 | 17 | 2194 | ||
XM_005258809.2 | XP_005258866.1 | 17 | 2194 | ||
XM_011526837.1 | XP_011525139.1 | 17 | 2179 | ||
XM_011526838.1 | XP_011525140.1 | 17 | 2194 | ||
XM_011526839.1 | XP_011525141.1 | 15 | 1843 | ||
XM_011526840.2 | XP_011525142.1 | 10 | 1189 | ||
XM_011526841.2 | XP_011525143.1 | 8 | 810 | ||
XM_011526842.1 | XP_011525144.1 | 7 | 642 | ||
XM_011526844.2 | XP_011525146.1 | 1 | 30 | ||
XM_017026665.1 | XP_016882154.1 | 17 | 2194 | ||
GenBank mRNA | BC058939.1 | - | 17 | 2138 | |
BQ880028.1 | - | 4 | 363 | ||
BQ960310.1 | - | 7 | 692 | ||
BX647726.1 | - | 18 | 2255 |