Gene Transcripts

Gene Symbol
Cep95
Entrez Gene ID
287766
Gene Name
centrosomal protein 95kDa
Gene Aliases
Ccdc45, RGD1309655
Chromosome Location
Chr. 10: 94988362 - 95016347 on Build Rnor_6.0
UniGene ID
Rn.155284
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013862.1 NP_001013884.1 15 2088
GenBank mRNA BC083896.1 - 15 2088

View Full Product Details