Gene Transcripts

Gene Symbol
Thnsl1
Entrez Gene ID
208967
Gene Name
threonine synthase-like 1 (bacterial)
Gene Aliases
AW413632
Chromosome Location
Chr. 2: 21205385 - 21215009 on Build GRCm38
UniGene ID
Mm.268841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001297.2 NP_001001297.1 3 417
NM_177588.2 NP_808256.1 3 1989
GenBank mRNA AK029518.1 - 3 1990
AK033465.1 - 3 386
AK083895.1 - 3 1989
AK087148.1 - 3 499
BC027743.1 - 1 514
BC090667.1 - 1 1698 30686917
BC117854.2 - 2 1773 40094242

View Full Product Details