Gene Transcripts

Gene Symbol
Wasf2
Entrez Gene ID
313024
Gene Name
WAS protein family, member 2
Gene Aliases
Chromosome Location
Chr. 5: 151319382 - 151385051 on Build Rnor_6.0
UniGene ID
Rn.162111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013167.1 NP_001013185.1 3 434
XM_008764138.1 XP_008762360.1 4 615
GenBank mRNA BC089207.1 - 3 434

View Full Product Details