Gene Transcripts

Gene Symbol
Tex29
Entrez Gene ID
498664
Gene Name
testis expressed 29
Gene Aliases
Chromosome Location
Chr. 16: 82985834 - 83006555 on Build Rnor_6.0
UniGene ID
Rn.195390
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017508.1 NP_001017508.1 6 1187
XM_008771406.1 XP_008769628.1 6 7915
GenBank mRNA BC079268.1 - 6 1187

View Full Product Details