Gene Transcripts
- Gene Symbol
- TMEM144
- Entrez Gene ID
- 55314
- Gene Name
- transmembrane protein 144
- Gene Aliases
- Chromosome Location
- Chr. 4: 158201597 - 158255287 on Build GRCh38
- UniGene ID
- Hs.176227
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018342.4 |
NP_060812.2
|
4, 5 |
760 |
|
XM_005263110.1 |
XP_005263167.1
|
5, 6 |
715 |
|
XM_005263112.3 |
XP_005263169.1
|
6, 7 |
750 |
|
XM_006714254.1 |
XP_006714317.1
|
5, 6 |
572 |
|
XM_017008366.1 |
XP_016863855.1
|
3, 4 |
728 |
|
GenBank mRNA |
AK002017.1 |
- |
3, 4 |
664 |
|
BC026318.1 |
- |
4, 5 |
454 |
|
BC036551.1 |
- |
3, 4 |
2038 |
|
BC048117.1 |
- |
3, 4 |
2033 |
|
BC054487.1 |
- |
4, 5 |
508 |
5732836 |
BC060875.1 |
- |
4, 5 |
486 |
4800219 |
View Full Product Details