Gene Transcripts
- Gene Symbol
- Kcnt1
- Entrez Gene ID
- 227632
- Gene Name
- potassium channel, subfamily T, member 1
- Gene Aliases
- C030030G16Rik, Slack, Slo2.2, slo2
- Chromosome Location
- Chr. 2: 25863791 - 25918273 on Build GRCm38
- UniGene ID
- Mm.119462
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK048406.1 |
- |
14 |
3193 |
|
CD363556.1 |
- |
1 |
319 |
|
View Full Product Details