Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289010.1 | NP_001275939.1 | 1, 2 | 875 | |
NM_020379.3 | NP_065112.1 | 1, 2 | 875 | ||
XM_017001860.1 | XP_016857349.1 | 1, 2 | 94 | ||
XM_017001861.1 | XP_016857350.1 | 1, 2 | 1523 | ||
GenBank mRNA | AB209275.1 | - | 1, 2 | 223 | |
AF261655.1 | - | 1, 2 | 875 | ||
BC137017.1 | - | 1, 2 | 681 | 9021010 |