Gene Transcripts

Gene Symbol
Olfr412
Entrez Gene ID
258153
Gene Name
olfactory receptor 412
Gene Aliases
MOR127-5P
Chromosome Location
Chr. 11: 74364671 - 74365609 on Build GRCm38
UniGene ID
Mm.376124
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011851.1 NP_001011851.1 1 97

View Full Product Details