Gene Transcripts

Gene Symbol
Olfr457
Entrez Gene ID
258989
Gene Name
olfactory receptor 457
Gene Aliases
MOR257-1
Chromosome Location
Chr. 6: 42471235 - 42472176 on Build GRCm38
UniGene ID
Mm.377778
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146987.1 NP_667198.1 1 815
GenBank mRNA BC119371.1 - 1 839
BC119373.1 - 1 839 8734122

View Full Product Details