Gene Transcripts

Gene Symbol
As3mt
Entrez Gene ID
140925
Gene Name
arsenite methyltransferase
Gene Aliases
Cyt19
Chromosome Location
Chr. 1: 266482858 - 266514570 on Build Rnor_6.0
UniGene ID
Rn.95453
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080890.1 NP_543166.1 4 332
GenBank mRNA AF393243.1 - 4 332
BC061533.1 - 4 356 6918024

View Full Product Details