Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008175.4 | NP_032201.2 | 12 | 1719 | |
GenBank mRNA | AK013678.1 | - | 12 | 1535 | |
AK018744.1 | - | 12 | 1560 | ||
AK147932.1 | - | 12 | 1536 | ||
AK149570.1 | - | 12 | 1537 | ||
AK150205.1 | - | 12 | 1535 | ||
AK150338.1 | - | 12 | 1524 | ||
AK150420.1 | - | 12 | 1523 | ||
AK150717.1 | - | 12 | 1535 | ||
AK150746.1 | - | 12 | 1537 | ||
AK150881.1 | - | 12 | 1537 | ||
AK151007.1 | - | 12 | 1535 | ||
AK151162.1 | - | 12 | 1537 | ||
AK151343.1 | - | 12 | 1535 | ||
AK151713.1 | - | 12 | 1535 | ||
AK151756.1 | - | 12 | 1535 | ||
AK152048.1 | - | 12 | 1535 | ||
AK152087.1 | - | 12 | 1535 | ||
AK152118.1 | - | 12 | 1524 | ||
AK152213.1 | - | 12 | 1524 | ||
AK152705.1 | - | 12 | 1535 | ||
AK152765.1 | - | 12 | 1535 | ||
AK152944.1 | - | 12 | 1524 | ||
AK153183.1 | - | 12 | 1525 | ||
AK153269.1 | - | 12 | 1534 | ||
AK153289.1 | - | 12 | 1535 | ||
AK153418.1 | - | 12 | 1535 | ||
AK153470.1 | - | 12 | 1535 | ||
AK154492.1 | - | 12 | 1535 | ||
AK159397.1 | - | 12 | 1524 | ||
AK159428.1 | - | 12 | 1524 | ||
AK159557.1 | - | 12 | 1535 | ||
AK159809.1 | - | 12 | 1535 | ||
AK160022.1 | - | 12 | 1524 | ||
AK160033.1 | - | 12 | 1535 | ||
AK160074.1 | - | 12 | 1524 | ||
AK185467.1 | - | 1 | 48 | ||
BC037047.1 | - | 11 | 1516 | ||
BC049943.1 | - | 12 | 1521 | ||
M86736.1 | - | 12 | 1517 | ||
X62321.1 | - | 11 | 1500 |