Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008973.2 | NP_032999.1 | 3 | 1601 | |
GenBank mRNA | AK011346.1 | - | 3 | 458 | |
AK011699.1 | - | 3 | 384 | ||
AK017768.1 | - | 3 | 420 | ||
AK133835.1 | - | 3 | 401 | ||
AK161847.1 | - | 3 | 351 | ||
AK161964.1 | - | 3 | 291 | ||
BC002064.1 | - | 3 | 170 | 3488877 | |
BC048363.1 | - | 3 | 175 | ||
BC061695.1 | - | 3 | 401 | 6313227 | |
D90225.1 | - | 3 | 345 |