Gene Transcripts

Gene Symbol
Tspyl1
Entrez Gene ID
22110
Gene Name
testis-specific protein, Y-encoded-like 1
Gene Aliases
Tspyl
Chromosome Location
Chr. 10: 34282190 - 34284885 on Build GRCm38
UniGene ID
Mm.20445
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009433.3 NP_033459.1 1 830
GenBank mRNA AF042180.1 - 1 794
AK053812.1 - 1 781
AK159432.1 - 1 811
AK166438.1 - 1 811
AK166805.1 - 1 759
AK166893.1 - 1 759
BC011213.1 - 1 777 4196841

View Full Product Details