Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136060.2 | NP_001129532.1 | 4 | 654 | |
NM_001198886.1 | NP_001185815.1 | 4 | 738 | ||
NM_010075.2 | NP_034205.1 | 4 | 989 | ||
NM_207282.3 | NP_997165.2 | 4 | 842 | ||
GenBank mRNA | AF092507.1 | - | 4 | 997 | |
AK031903.1 | - | 4 | 615 | ||
AK051708.1 | - | 4 | 820 | ||
AK082186.1 | - | 4 | 913 | ||
AK158964.1 | - | 4 | 843 | ||
BC048383.1 | - | 4 | 505 | 6306267 | |
BC085154.1 | - | 4 | 738 | 6838230 |