Gene Transcripts
- Gene Symbol
- Cyth2
- Entrez Gene ID
- 19158
- Gene Name
- cytohesin 2
- Gene Aliases
- ARNO, CLM2, Pscd2
- Chromosome Location
- Chr. 7: 45806637 - 45814316 on Build GRCm38
- UniGene ID
- Mm.272130
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001112701.1 |
NP_001106171.1
|
4, 5 |
561 |
|
NM_011181.3 |
NP_035311.1
|
4, 5 |
561 |
|
GenBank mRNA |
AB013466.1 |
- |
4, 5 |
496 |
|
AB013467.1 |
- |
3, 4 |
1064 |
|
AF079971.1 |
- |
4, 5 |
530 |
|
AK045451.1 |
- |
4, 5 |
530 |
|
AK078067.1 |
- |
4, 5 |
557 |
|
AK143829.1 |
- |
4, 5 |
503 |
|
AK168979.1 |
- |
4, 5 |
501 |
|
AK172050.1 |
- |
4, 5 |
517 |
|
BC004662.1 |
- |
4, 5 |
475 |
3498490 |
BC088833.1 |
- |
4, 5 |
535 |
5293465 |
View Full Product Details