Gene Transcripts

Gene Symbol
Scarb1
Entrez Gene ID
20778
Gene Name
scavenger receptor class B, member 1
Gene Aliases
AI120173, CD36, Cd36l1, Cla-1, Cla1, D5Ertd460e, Hdlq1, Hlb398, SR-B1, SR-BI, SRBI, Srb1, mSR-BI
Chromosome Location
Chr. 5: 125277087 - 125341111 on Build GRCm38
UniGene ID
Mm.282242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205082.1 NP_001192011.1 2, 3 504
NM_001205083.1 NP_001192012.1 2, 3 504
NM_016741.2 NP_058021.1 2, 3 504
GenBank mRNA AK010622.1 - 2, 3 429
AK028191.1 - 2, 3 485
AK033114.1 - 2, 3 429
AK145588.1 - 2, 3 430
AK154933.1 - 2, 3 496
AK158122.1 - 2, 3 429
AK169906.1 - 2, 3 480
AK171602.1 - 2, 3 483
BC004656.1 - 2, 3 484 3498018
CT010222.1 - 2, 3 292
U37799.1 - 2, 3 342

View Full Product Details