Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004823.1 | NP_004814.1 | 2 | 478 | |
XM_011527526.1 | XP_011525828.1 | 3 | 1040 | ||
GenBank mRNA | AF117708.1 | - | 2 | 478 | |
AF134149.1 | - | 2 | 487 | ||
AF281302.1 | - | 2 | 443 | ||
AF281303.1 | - | 2 | 67 | ||
AK022344.1 | - | 2 | 207 | ||
AK222545.1 | - | 2 | 460 | ||
AK315653.1 | - | 2 | 452 | ||
BC004367.1 | - | 2 | 408 | 3639657 | |
EU978937.1 | - | 2 | 426 |