Gene Transcripts
- Gene Symbol
- KCNK6
- Entrez Gene ID
- 9424
- Gene Name
- potassium two pore domain channel subfamily K member 6
- Gene Aliases
- K2p6.1, KCNK8, TOSS, TWIK-2, TWIK2
- Chromosome Location
- Chr. 19: 38319814 - 38329014 on Build GRCh38
- UniGene ID
- Hs.240395
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004823.1 |
NP_004814.1
|
2, 3 |
823 |
|
XM_011527526.1 |
XP_011525828.1
|
3, 4 |
1385 |
|
GenBank mRNA |
AF117708.1 |
- |
2, 3 |
823 |
|
AF134149.1 |
- |
2, 3 |
832 |
|
AF281302.1 |
- |
2, 3 |
788 |
|
AF281303.1 |
- |
2, 3 |
412 |
|
AK022344.1 |
- |
2, 3 |
552 |
|
AK222545.1 |
- |
2, 3 |
805 |
|
AK315653.1 |
- |
2, 3 |
797 |
|
BC004367.1 |
- |
2, 3 |
753 |
3639657 |
EU978937.1 |
- |
2, 3 |
771 |
|
View Full Product Details