Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024216.1 | NP_077178.1 | 3, 4 | 444 | |
GenBank mRNA | AK016996.1 | - | 3, 4 | 375 | |
AK042405.1 | - | 1067 | |||
AK080720.1 | - | 3, 4 | 391 | ||
AK087968.1 | - | 3, 4 | 395 | ||
AK146783.1 | - | 3, 4 | 382 | ||
BC003341.1 | - | 3, 4 | 444 | 3257412 |