gRNA Design Details
- Target DNA Sequence
- CTCAGGTGATCGAGTTGGAG
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 23681503 - 23681525 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NKX3-1
- Entrez Gene ID
-
4824
- Gene Name
- NK3 homeobox 1
- Gene Aliases
-
BAPX2, NKX3, NKX3.1, NKX3A
- Chromosome Location
-
Chr.8: 23678693 - 23682937 on GRCh38
- UniGene ID
- Hs.55999
- Species
-
Human