gRNA Design Details
- Target DNA Sequence
- GTTTGACACAGAGCTGATCG
- PAM Sequence
- GGG
- Target locus
-
Chr.16: 68337502 - 68337524 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.87/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRMT7
- Entrez Gene ID
-
54496
- Gene Name
- protein arginine methyltransferase 7
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 68310974 - 68358832 on GRCh38
- UniGene ID
- Hs.640229
- Species
-
Human