gRNA Design Details

Target DNA Sequence
CTTTCACCTTTGATACGAAG
PAM Sequence
GGG
Target locus
Chr.11: 64841878 - 64841900 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.09/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CDC42BPG
Entrez Gene ID
55561
Gene Name
CDC42 binding protein kinase gamma
Gene Aliases
DMPK2, HSMDPKIN, KAPPA-200, MRCKG, MRCKgamma
Chromosome Location
Chr.11: 64823809 - 64844685 on GRCh38
UniGene ID
Hs.293590
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_017525.2 NP_059995.2 2
XM_011545155.2 XP_011543457.1 2
XM_011545156.2 XP_011543458.1 2
XM_011545157.2 XP_011543459.1 2
XM_011545158.2 XP_011543460.1 2
XM_011545159.2 XP_011543461.1 2
XM_011545160.2 XP_011543462.1 3
XM_011545161.2 XP_011543463.1 2
XM_017017996.1 XP_016873485.1 2
XM_017017997.1 XP_016873486.1 2
XM_017017998.1 XP_016873487.1 2