gRNA Design Details
- Target DNA Sequence
- AGCCACGGGTTTAAAACCTT
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 82496135 - 82496157 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMEM150C
- Entrez Gene ID
-
441027
- Gene Name
- transmembrane protein 150C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 82484451 - 82561973 on GRCh38
- UniGene ID
- Hs.507676
- Species
-
Human