gRNA Design Details
- Target DNA Sequence
- TGGCTGGCGAAGATAGTCCG
- PAM Sequence
- AGG
- Target locus
-
Chr.14: 24131789 - 24131811 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FITM1
- Entrez Gene ID
-
161247
- Gene Name
- fat storage inducing transmembrane protein 1
- Gene Aliases
-
FIT1
- Chromosome Location
-
Chr.14: 24131466 - 24132849 on GRCh38
- UniGene ID
- Hs.128060
- Species
-
Human