gRNA Design Details
- Target DNA Sequence
- CCTAGTGACTTAACTGCCGA
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 93758522 - 93758544 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMEM67
- Entrez Gene ID
-
91147
- Gene Name
- transmembrane protein 67
- Gene Aliases
-
JBTS6, MECKELIN, MKS3, NPHP11, TNEM67
- Chromosome Location
-
Chr.8: 93754844 - 93832653 on GRCh38
- UniGene ID
- Hs.116240
- Species
-
Human