gRNA Design Details
- Target DNA Sequence
- GAGCGGCATGTTGGACCATA
- PAM Sequence
- GGG
- Target locus
-
Chr.15: 29053914 - 29053936 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- APBA2
- Entrez Gene ID
-
321
- Gene Name
- amyloid beta precursor protein binding family A member 2
- Gene Aliases
-
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L
- Chromosome Location
-
Chr.15: 28885479 - 29118315 on GRCh38
- UniGene ID
- Hs.618112
- Species
-
Human