gRNA Design Details
- Target DNA Sequence
- GATCATGGCCGTGCCCATGA
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 99405875 - 99405897 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmbim6
- Entrez Gene ID
-
110213
- Gene Name
- transmembrane BAX inhibitor motif containing 6
- Gene Aliases
-
5031406P05Rik, Bi1, Tegt
- Chromosome Location
-
Chr.15: 99392947 - 99410049 on GRCm38
- UniGene ID
- Mm.16769
- Species
-
Mouse