gRNA Design Details
- Target DNA Sequence
- GACTCTTTAACGCTTGCCGT
- PAM Sequence
- GGG
- Target locus
-
Chr.14: 93185439 - 93185461 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMEM251
- Entrez Gene ID
-
26175
- Gene Name
- transmembrane protein 251
- Gene Aliases
-
C14orf109
- Chromosome Location
-
Chr.14: 93184951 - 93187086 on GRCh38
- UniGene ID
- Hs.275352
- Species
-
Human