gRNA Design Details
- Target DNA Sequence
- GTAGTTATCCAAGAGGACCT
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 47348579 - 47348601 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.63/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RBP3
- Entrez Gene ID
-
5949
- Gene Name
- retinol binding protein 3
- Gene Aliases
-
D10S64, D10S65, D10S66, IRBP, RBPI, RP66
- Chromosome Location
-
Chr.10: 47348371 - 47357875 on GRCh38
- UniGene ID
- Hs.591928
- Species
-
Human