gRNA Design Details
- Target DNA Sequence
- CCTCTCTATCGTGTACGCTT
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 154429015 - 154429037 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
94.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPR149
- Entrez Gene ID
-
344758
- Gene Name
- G protein-coupled receptor 149
- Gene Aliases
-
IEDA, PGR10
- Chromosome Location
-
Chr.3: 154337672 - 154429715 on GRCh38
- UniGene ID
- Hs.688231
- Species
-
Human